1-Heptanol for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

8.20624.0500 8.20624.1000 8.20624.0100
8.20624.0500EA 52.9 GBP
8.20624.0500 8.20624.1000 8.20624.0100
1-Heptanol for synthesis, Sigma-Aldrich®
1-Heptanol
Formula: H₃C(CH₂)₅CH₂OH
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002986
CAS Number: 111-70-6
EINECS: 203-897-9
Merck Index: 13,04681

Order Now

Specification Test Results

Solubility 1 g/l (18 °C)
Melting Point -35 °C
Molar Mass 116.2 g/mol
Boiling Point 176 °C (1013 hPa)
Vapor Pressure 0.5 hPa (20 °C)
Flash Point 78 °C
Refractive Index 1.4242 (20 °C, 589 nm)
Explosion Limit 0.9 %(V)
Density 0.82 g/cm3 (20 °C)
Ignition Point 350 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR