1-Dodecanethiol for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Mercaptan C12, 1-Mercaptododecane, n-Dodecanethiol, Lauryl mercaptan, n-Dodecyl mercaptan, NDM

M-Clarity™ quality level = MQ200

8.20544.0250 8.20544.1000 8.20544.9011 8.20544.9170
8.20544.0250EA 50.9 GBP
8.20544.0250 8.20544.1000 8.20544.9011 8.20544.9170
1-Dodecanethiol for synthesis, Sigma-Aldrich®
1-Dodecanethiol
Formula: C₁₂H₂₆S
Boiling Pt: 269…273 °C (1013 hPa)
Melting Pt: –7 °C
Density: 0.855 g/cm³ (20 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00004885
CAS Number: 112-55-0
EINECS: 203-984-1
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Solubility  - 1 g/l (22 °C)
Melting Point -7 °C
Molar Mass 202.39 g/mol
Boiling Point 275 °C (1013 hPa)
Vapor Pressure 0.002 hPa (20 °C)
Flash Point 127 °C
Refractive Index 1.4587 (20 °C, 589 nm)
Density 0.845 g/cm3 (20 °C)
Ignition Point 230 °C
Synonyms Lauryl mercaptan, Dodecyl mercaptan
Hill Formula C12H26S
Chemical Formula CH3(CH2)11SH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR