meso-Hydrobenzoin for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.20526.0001
8.20526.0001EA 54.2 GBP
8.20526.0001
meso-Hydrobenzoin for synthesis, Sigma-Aldrich®
meso-Hydrobenzoin
Formula: C₁₄H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00004499
CAS Number: 492-70-6
EINECS: 207-758-3

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 128 - 131 °C
Molar Mass 214.26 g/mol
Synonyms meso-Hydrobenzoin, meso-1,2-Diphenylethylene glycol
Hill Formula C14H14O2
Chemical Formula HOCH(C6H5)CH(C6H5)OH
Storage temperature +15...+25 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR