N,N-Dimethyl-4-nitrosoaniline for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.20515.0025
8.20515.0025EA 185 GBP
8.20515.0025
N,N-Dimethyl-4-nitrosoaniline for synthesis, Sigma-Aldrich®
N,N-Dimethyl-4-nitrosoaniline
Formula: C₈H₁₀N₂O
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 87 °C
Density: 1.145 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002063
CAS Number: 138-89-6
EINECS: 205-343-1
UN: 1369
ADR: 4.2,II
Merck Index: 12,06736

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 84 - 87 °C
Molar Mass 150.18 g/mol
Density 1.15 g/cm3 (20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR