(±)-Ethyl-2,3-dibromopropionate

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ethyl 2,3-Dibromopropionate

M-Clarity™ quality level = MQ200

8.20418.0100
8.20418.0100EA 59.8 GBP
8.20418.0100
(±)-Ethyl-2,3-dibromopropionate
(±)-Ethyl 2,3-dibromopropionate
Formula: C₅H₈Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00000212
CAS Number: 3674-13-3
EINECS: 222-941-8
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 259.92 g/mol
Boiling Point 211 - 214 °C (992 hPa)
Flash Point 91 °C
Refractive Index 1.4986 (20 °C, 589 nm)
Density 1.79 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR