Ethylamine 70% ≥70% in aqueous solution for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Monoethylamine, Aminoethane, Ethanamine

M-Clarity™ quality level = MQ200

8.20024.1000 8.20024.2500
8.20024.1000EA 39.6 GBP
8.20024.1000 8.20024.2500
Ethylamine 70% ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Ethylamine
Formula: C₂H₇N
Storage Temperature: Ambient
MDL Number: MFCD00008160
CAS Number: 75-04-7
EINECS: 200-834-7
UN: 2270
ADR: 3 (8),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Boiling Point 39 °C (1013 hPa)
Vapor Pressure 460 hPa (20 °C)
Flash Point -26 °C
Explosion Limit 3.0 - 12.8 %(V)
Density 0.81 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)
Ignition Point 375 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR