Tetrabutylammonium hydroxide 20% aqueous solution zS, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


MQ200

8.18759.0100 8.18759.0500 8.18759.0050
8.18759.0100EA 48.3 GBP
8.18759.0100 8.18759.0500 8.18759.0050
Tetrabutylammonium hydroxide 20% aqueous solution zS, Sigma-Aldrich®
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Melting Pt: 29.85
Density: 0.968
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Boiling Point 102 °C (1013 hPa)
Density 0.99 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR