4-Methoxythiophenol

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

8.18743.0005
8.18743.0005EA 0 GBP
8.18743.0005
4-Methoxythiophenol
4-Methoxythiophenol
Formula: C₇H₈OS
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 42 °C
Density: 1.1397 g/cm³ (20 °C)
MDL Number: MFCD00004849
CAS Number: 696-63-9
EINECS: 211-799-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR