Nicotinic acid (Niacin) for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

8.18714.0100 8.18714.1000
8.18714.0100EA 29.7 GBP
8.18714.0100 8.18714.1000
Nicotinic acid (Niacin) for synthesis, Sigma-Aldrich®
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 142 °C
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552

Order Now

Specification Test Results

Solubility 18 g/l (20 °C)
Melting Point 236.6 °C
Molar Mass 123.11 g/mol
Bulk Density 330 kg/m3
Flash Point 193 °C
Density 1.47 g/cm3 (20 °C)
pH 2.7 (18 g/l, H2O, 20 °C)
Ignition Point 365 °C Dust

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR