(1R)-(+)-α-Pinene for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (+)-α-Pinene, D(+)-α-Pinene, (1R)-(+)-2-Pinene, (+)-Pin-2(3)-ene

M-Clarity™ quality level = MQ200

8.18632.0100
8.18632.0100EA 70 GBP
8.18632.0100
(1R)-(+)-α-Pinene for synthesis, Sigma-Aldrich®
(1R)-(+)-α-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.857 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001346
CAS Number: 7785-70-8
EINECS: 232-087-8
UN: 2368
ADR: 3,III
Merck Index: 12,07599

Order Now

Specification Test Results

Melting Point -62 °C
Molar Mass 136.23 g/mol
Boiling Point 155 - 156 °C (1013 hPa)
Vapor Pressure 5 hPa (25 °C)
Flash Point 33 °C
Refractive Index 1.4657 (20 °C, 589 nm)
Explosion Limit 0.8 - 6 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 255 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR