(4-Carboxybutyl)triphenylphosphonium bromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

8.18330.0050
8.18330.0050EA 76.3 GBP
8.18330.0050
(4-Carboxybutyl)triphenylphosphonium bromide, Sigma-Aldrich®
(4-Carboxybutyl)triphenylphosphonium bromide
Formula: C₂₃H₂₄BrO₂P
Melting Pt: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00011906
CAS Number: 17814-85-6
EINECS: 241-782-5
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 204 - 208 °C
Molar Mass 443.31 g/mol
Bulk Density 360 - 400 kg/m3
Flash Point 195 °C
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR