(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (R)-2-Isopropyl-3,6-dimethoxy-2,5-dihydropyrazine, (2R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, (R)-3-Isopropyl-2,5-dimethoxy-3,6-dihydropyrazine

M-Clarity™ quality level = MQ200

8.18315.0100
8.18315.0100EA 0 GBP
8.18315.0100
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine for synthesis, Sigma-Aldrich®
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine
Formula: C₉H₁₆N₂O₂
MDL Number: MFCD00040565
CAS Number: 109838-85-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 184.23 g/mol
Boiling Point 100 - 102 °C (20 hPa)
Flash Point 85 °C
Density 1.03 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR