2-(Allylthio)-2-thiazoline for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.18083.0010
8.18083.0010EA 96.7 GBP
8.18083.0010
2-(Allylthio)-2-thiazoline for synthesis, Sigma-Aldrich®
2-(Allylthio)-2-thiazoline
Formula: C₄H₇NS
Storage Temperature: Ambient
CAS Number: 3571-74-2
EINECS: 222-679-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 159.28 g/mol
Boiling Point 92 - 94 °C (2 hPa)
Density 1.16 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR