L(+)-Aspartic acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D

M-Clarity™ quality level = MQ200

8.16003.0100 8.16003.0500
8.16003.0100EA 33.3 GBP
8.16003.0100 8.16003.0500
L(+)-Aspartic acid for synthesis, Sigma-Aldrich®
L(+)-Aspartic acid
Formula: C₄H₇NO₄
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 251 °C
Density: 1.6613 g/cm³ (13 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845

Order Now

Specification Test Results

Solubility 4 g/l (20 °C)
Melting Point 269 - 271 °C (decomposition)
Molar Mass 133.1 g/mol
Bulk Density 430 kg/m3
pH 2.5 - 3.5 (4 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR