4-Phenylmorpholine for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N-Phenylmorpholine

M-Clarity™ quality level = MQ200

8.14859.0050 8.14859.0250
8.14859.0050EA 46.7 GBP
8.14859.0050 8.14859.0250
4-Phenylmorpholine for synthesis, Sigma-Aldrich®
4-Phenylmorpholine
Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
Boiling Pt: 268
Melting Pt: 51…54 °C
Density: 1.058 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006166
CAS Number: 92-53-5
EINECS: 202-164-0
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Melting Point 48 - 51 °C
Molar Mass 163.22 g/mol
Boiling Point 260 °C (1013 hPa)
Flash Point 124 °C
Density 1.053 g/cm3 (60 °C)
Ignition Point 410 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR