Sodium chlorite 25% in aqueous solution for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.14815.0250 8.14815.1000
8.14815.0250EA 41 GBP
8.14815.0250 8.14815.1000
Sodium chlorite 25% in aqueous solution for synthesis, Sigma-Aldrich®
Sodium chlorite
Formula: ClNaO₂
Storage Temperature: Ambient
MDL Number: MFCD00003478
CAS Number: 7758-19-2
EINECS: 231-836-6
UN: 1908
ADR: 8,II

Order Now

Specification Test Results

Boiling Point 100 °C
Density 1.21 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR