Magnesium chloride for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.14733.0100 8.14733.0500 8.14733.9050
8.14733.0100EA 30.7 GBP
8.14733.0100 8.14733.0500 8.14733.9050
Magnesium chloride for synthesis, Sigma-Aldrich®
Magnesium chloride
Formula: MgCl₂
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 715 °C
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Solubility 542 g/l (20 °C)
Melting Point 712 °C
Molar Mass 95.22 g/mol
Boiling Point 1412 °C (1013 hPa)
Density 2.32 g/cm3 (20 °C)
pH =7 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR