2-Mercaptobenzothiazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT

M-Clarity™ quality level = MQ200

8.14674.0250 8.14674.1000
8.14674.0250EA 46.6 GBP
8.14674.0250 8.14674.1000
2-Mercaptobenzothiazole, Sigma-Aldrich®
2-Mercaptobenzothiazole
Formula: C₇H₅NS₂
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 177…182 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005781
CAS Number: 149-30-4
EINECS: 205-736-8
UN: 3077
ADR: 9,III
Merck Index: 13,05893

Order Now

Specification Test Results

Solubility 0.12 g/l (25 °C)
Melting Point 180 - 183 °C
Molar Mass 167.25 g/mol
Bulk Density 390 kg/m3
Boiling Point 260 °C decomposes
Vapor Pressure  - 0.000003 hPa (25 °C)
Flash Point 243 °C
Explosion Limit 15 %(V)
Density 1.42 g/cm3 (20 °C)
pH 7 (0.12 g/l, H2O, 25 °C)
Ignition Point 465 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR