L(+)-Glutamic acid hydrochloride for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: (S)-2-Aminopentanedioic acid hydrochloride, 2-Aminoglutaric acid hydrochloride

M-Clarity™ quality level = MQ200

8.14632.0250 8.14632.9025
8.14632.0250EA 33.5 GBP
8.14632.0250 8.14632.9025
L(+)-Glutamic acid hydrochloride for synthesis, Sigma-Aldrich®
L(+)-Glutamic acid hydrochloride
Formula: C₅H₁₀ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD00012619
CAS Number: 138-15-8
EINECS: 205-315-9

Order Now

Specification Test Results

Solubility 490 g/l (20 °C)
Melting Point 191 - 193 °C decomposes
Molar Mass 183.59 g/mol
Bulk Density 630 kg/m3
Density 1.525 g/cm3

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR