3-Methoxy-3-methyl-1-butanol for synthesis

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

8.14337.0100 8.14337.0500 8.14337.9025 8.14337.9185
8.14337.0100EA 40.2 GBP
8.14337.0100 8.14337.0500 8.14337.9025 8.14337.9185
3-Methoxy-3-methyl-1-butanol for synthesis
3-Methoxy-3-methyl-1-butanol
Formula: C₆H₁₄O₂
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.9038 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044771
CAS Number: 56539-66-3
EINECS: 260-252-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR