Imidazole for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.14223.0005 8.14223.0250 8.14223.1000 8.14223.9025
8.14223.0005EA 23.4 GBP
8.14223.0005 8.14223.0250 8.14223.1000 8.14223.9025
Imidazole for synthesis, Sigma-Aldrich®
Imidazole
Formula: C₃H₄N₂
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 89…90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8,III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 633 g/l (20 °C)
Melting Point 89 - 90 °C
Molar Mass 68.08 g/mol
Bulk Density 500 - 600 kg/m3
Boiling Point 256 °C (1013 hPa)
Vapor Pressure 0.003 hPa (20 °C)
Flash Point 145 °C
Density 1.030 g/cm3
pH 10.5 (67 g/l, H2O, 20 °C)
Ignition Point 480 °C
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR