4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 99 GC_ASSAY_METHOD zS, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Cryptand 2.2.2, Fludeoxyglucose Related Compound A, Kryptofix 222, 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo(8.8.8)hexacosane, 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane, Cryptand 222, (2.2.2)Cryptand

MQ200

8.10647.0001 8.10647.0050 8.10647.0005 8.10647.0500 8.10647.8250 8.10647.1000
8.10647.0001EA 136 GBP
8.10647.0001 8.10647.0050 8.10647.0005 8.10647.0500 8.10647.8250 8.10647.1000
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 99 GC_ASSAY_METHOD zS, Sigma-Aldrich®
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)
Formula: C₁₈H₃₆N₂O₆
MW: 376.49 g/mol
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00005111
CAS Number: 23978-09-8
EINECS: 245-962-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a74ac532561e4fecf111f3260b402bcd1fb0af9&vid=13491901da172b5971e6d7f330c893460a77cd23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) ≥ 99
Identity (IR-spectrum) passes test
Melting range (°C) 70 - 73


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR