Tributylamine for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.08358.0100 8.08358.1000 8.08358.2500 8.08358.9040
8.08358.0100EA 31.2 GBP
8.08358.0100 8.08358.1000 8.08358.2500 8.08358.9040
Tributylamine for synthesis, Sigma-Aldrich®
Tributylamine
Formula: C₁₂H₂₇N
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.7771 g/cm³ (20 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00009431
CAS Number: 102-82-9
EINECS: 203-058-7
UN: 2542
ADR: 6.1,II
Merck Index: 13,09691

Order Now

Specification Test Results

Solubility 0.3 g/l (20 °C)
Melting Point -70 °C
Molar Mass 185.35 g/mol
Boiling Point 216 - 217 °C (1013 hPa)
Vapor Pressure 0.18 hPa (20 °C)
Flash Point 63 - 66 °C
Explosion Limit 1.4 - 6.0 %(V)
Density 0.78 g/cm3 (20 °C)
pH 10.6 (H2O, 20 °C) (saturated aqueous solution)
Ignition Point 190 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR