Dimethyl terephthalate for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.08177.0100 8.08177.1000 8.08177.2500
8.08177.0100EA 25.7 GBP
8.08177.0100 8.08177.1000 8.08177.2500
Dimethyl terephthalate for synthesis, Sigma-Aldrich®
Dimethyl terephthalate
Formula: C₁₀H₁₀O₄
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: 139 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 141 °C
Storage Temperature: Ambient
MDL Number: MFCD00008440
CAS Number: 120-61-6
EINECS: 204-411-8
Merck Index: 12,09306

Order Now

Specification Test Results

Solubility 0.036 g/l (20 °C)
Melting Point 139 - 141 °C
Molar Mass 194.19 g/mol
Bulk Density 500 kg/m3
Boiling Point 282 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 154 °C (External MSDS)
Explosion Limit 0.8 - 11.8 %(V)
Density 1.35 g/cm3 (20 °C)
Ignition Point 520 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR