Pyromellitic dianhydride for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.07516.0100
8.07516.0100EA 31.6 GBP
8.07516.0100
Pyromellitic dianhydride for synthesis, Sigma-Aldrich®
Pyromellitic dianhydride
Formula: C₁₀H₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00005005
CAS Number: 89-32-7
EINECS: 201-898-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR