Propargyl alcohol for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Propyn-1-ol, Prop-2-yn-1-ol

M-Clarity™ quality level = MQ200

8.07050.0250 8.07050.1000
8.07050.0250EA 37.1 GBP
8.07050.0250 8.07050.1000
Propargyl alcohol for synthesis, Sigma-Aldrich®
Propargyl alcohol
Formula: C₃H₄O
Boiling Pt: 114 °C (1013 hPa)
Melting Pt: –53 °C
Density: 0.9485 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00002912
CAS Number: 107-19-7
EINECS: 203-471-2
UN: 2929
ADR: 6.1 (3),II
Merck Index: 12,07993

Order Now

Specification Test Results

Solubility 1000 g/l (20 °C)
Melting Point -53 °C
Molar Mass 56.06 g/mol
Boiling Point 114 - 115 °C
Vapor Pressure 10 hPa (20 °C)
Flash Point 33.5 °C
Refractive Index 1.4318 (20 °C, 589 nm)
Explosion Limit 1.9 - 86.2 %(V)
Density 0.95 g/cm3 (20 °C)
pH 3 (500 g/l, H2O, 20 °C)
Ignition Point 365 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR