Phenyl isothiocyanate for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.07028.0100 8.07028.0250 8.07028.0025
8.07028.0100EA 71.6 GBP
8.07028.0100 8.07028.0250 8.07028.0025
Phenyl isothiocyanate for synthesis, Sigma-Aldrich®
Phenyl isothiocyanate
Formula: C₇H₅NS
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: –21 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004798
CAS Number: 103-72-0
EINECS: 203-138-1
UN: 2206
ADR: 6.1,II
Merck Index: 12,07450

Order Now

Specification Test Results

Solubility (20 °C) insoluble,(decomposition)
Melting Point -21 °C
Molar Mass 135.18 g/mol
Boiling Point 221 °C (1013 hPa)
Vapor Pressure 10 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.6497 (20 °C, 589 nm)
Density 1.13 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Synonyms Isothiocyanic acid phenyl ester, PTC
Hill Formula C7H5NS
Chemical Formula C6H5NCS

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR