Nickel(II) acetylacetonate for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.06715.0025 8.06715.0100 8.06715.9025
8.06715.0025EA 46.2 GBP
8.06715.0025 8.06715.0100 8.06715.9025
Nickel(II) acetylacetonate for synthesis, Sigma-Aldrich®
Nickel (II) acetylacetonate
Formula: C₁₀H₁₄NiO₄
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 208 °C
Density: 1.455 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000024
CAS Number: 3264-82-2
EINECS: 221-875-7
Merck Index: 12,06584

Order Now

Specification Test Results

Solubility 4.8 g/l (20 °C)
Melting Point 208 °C
Molar Mass 256.91 g/mol
Bulk Density 470 kg/m3
Boiling Point 220 - 235 °C (14 hPa)
Vapor Pressure 2.7 hPa (110 °C)
Synonyms Bis(acetylacetonato)nickel(II), Bis(2,4-pentanedionato) nickel(II)
Hill Formula C10H14NiO4

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR