1-Chlorododecane for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ200

8.05340.0100 8.05340.1000
8.05340.0100EA 31.2 GBP
8.05340.0100 8.05340.1000
1-Chlorododecane for synthesis, Sigma-Aldrich®
1-Chlorododecane
Formula: C₁₂H₂₅Cl
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: –9 °C
Density: 0.867 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013688
CAS Number: 112-52-7
EINECS: 203-981-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility  - 0.007 g/l (25 °C)
Melting Point -9 °C
Molar Mass 204.78 g/mol
Boiling Point 124 °C (13 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 130 °C
Density 0.87 g/cm3 (20 °C)
Ignition Point 190 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR