Glutardialdehyde bis(sodium hydrogen sulphite) for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ200

8.04240.0250
8.04240.0250EA 54.8 GBP
8.04240.0250
Glutardialdehyde bis(sodium hydrogen sulphite) for synthesis, Sigma-Aldrich®
Glutardialdehyde bis(sodium hydrogen sulphite)
Formula: C₅H₁₀Na₂O₈S₂
MW: 308.24 g/mol
CAS Number: 7420-89-5
EINECS: 231-043-5

Order Now

Specification Test Results

Solubility 285 g/l (20 °C)
Molar Mass 308.22 g/mol
pH 3 - 5 (100 g/l, H2O, 20 °C)
Synonyms Pentane-1,5-dial-bis(sodium bisulfit)
Hill Formula C5H10Na2O8S2
Chemical Formula CH2[CH2CH(OH)SO3Na]2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR