N,N-Dimethylethylendiamine for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N,N-Dimethylethylenediamine, asym. Dimethylethylendiamine, unsym-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, 2-Dimethylaminoethylamine, 2-Aminoethyldimethylamine

M-Clarity™ quality level = MQ200

8.03779.0100 8.03779.9022
8.03779.0100EA 57.1 GBP
8.03779.0100 8.03779.9022
N,N-Dimethylethylendiamine for synthesis, Sigma-Aldrich®
N,N-Dimethylethylendiamine
Formula: C₄H₁₂N₂
Boiling Pt: ∼106 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.804 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008175
CAS Number: 108-00-9
EINECS: 203-541-2
UN: 2733
ADR: 3 (8),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 88.15 g/mol
Boiling Point 107 - 109 °C (1013 hPa)
Vapor Pressure 29 hPa (20 °C)
Flash Point 12.5 °C
Explosion Limit 1.3 - 10.8 %(V)
Density 0.81 g/cm3 (20 °C)
pH 11.4 (10 g/l, H2O, 20 °C)
Ignition Point 225 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR