Divinylbenzene (mixture of isomers) stabilised, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: DVB, Divinylbenzene

Stabilisation: Stabilised with 4-tert-butylpyrocatechol (TBC)
M-Clarity™ quality level = MQ200

8.03598.0250 8.03598.1000 8.03598.6010
8.03598.0250EA 48.1 GBP
8.03598.0250 8.03598.1000 8.03598.6010
Divinylbenzene (mixture of isomers) stabilised, Sigma-Aldrich®
Divinylbenzene (mixture of isomers)
Formula: C₁₀H₁₀
Boiling Pt: 195 °C (1013 hPa)
Density: 0.91 g/cm3 (20 °C)
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01778656
CAS Number: 1321-74-0
EINECS: 215-325-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay isomers (GC, area%) 60.0 - 65.0 % (a/a)
Density (d 20 °C/ 4 °C) 0.913 - 0.916
Assay isomers ethylvinyl benzol (GC, area%) 34.0 - 39.0 % (a/a)
Identity (IR) passes test
The material is stabilised with 4-tert-Butylpyrocatechol. At time of analytical testing a content of >300 ppm stabiliser was determined. 0.9 hPa (20 °C)
Flash Point 77 °C
Explosion Limit 1.1 - 6.2 %(V)
Density 0.91 g/cm3 (20 °C)
Ignition Point 470 °C
Synonyms DVB
Hill Formula C10H10

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR