Diethylene glycol for synthesis

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 2,2'-Oxybis(ethan-1-ol), Digol, 2,2'-Oxybisethanol

8.03131.1000 8.03131.5000 8.03131.9025
8.03131.1000EA 27.1 GBP
8.03131.1000 8.03131.5000 8.03131.9025
Diethylene glycol for synthesis
Diethylene glycol
Formula: C₄H₁₀O₃
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 123 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2
Merck Index: 13,03146
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f023bd53ecff654f6d6d398b3ee2a1bac87d56f&vid=25d8943f9c247b4203b1e1bcba59bf77dfdd9158&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR