Phenyl chloroformate for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.02353.0250 8.02353.9010
8.02353.0250EA 57.1 GBP
8.02353.0250 8.02353.9010
Phenyl chloroformate for synthesis, Sigma-Aldrich®
Phenyl chloroformate
Formula: C₇H₅ClO₂
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: –28 °C
Density: 1.248 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00000637
CAS Number: 1885-14-9
EINECS: 217-547-8
UN: 2746
ADR: 6.1 (8),II

Order Now

Specification Test Results

Solubility (20 °C) decomposes,Hydrolysis
Melting Point -28 °C
Molar Mass 156.57 g/mol
Boiling Point 188 - 189 °C (1013 hPa)
Vapor Pressure 0.9 hPa (20 °C)
Flash Point 69 °C
Refractive Index 1.511 (20 °C, 589 nm)
Density 1.24 g/cm3 (20 °C)
Ignition Point 540 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR