Anisole zS, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Methyl phenyl ether, Methoxybenzene

MQ200

8.01452.0100 8.01452.0500 8.01452.1000 8.01452.2500 8.01452.9050 8.01452.9025
8.01452.0100EA 27.3 GBP
8.01452.0100 8.01452.0500 8.01452.1000 8.01452.2500 8.01452.9050 8.01452.9025
Anisole zS, Sigma-Aldrich®
Anisole
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 154
Melting Pt: –37 °C
Density: 0.9961 g/cm³ (20 °C)
Flash Pt: 44
Storage Temperature: Ambient
MDL Number: MFCD00008354
CAS Number: 100-66-3
EINECS: 202-876-1
UN: 2222
ADR: 3 III
Merck Index: 13,00672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d32206d4bb9dbd12360d11d39b31237d54a8dcf8&vid=286a07a5e34a1ec55ff6339785af95d9a4326825&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 1.6 g/l (25 °C)
Melting Point -37 °C
Molar Mass 108.13 g/mol
Boiling Point 156 °C (1013 hPa)
Vapor Pressure 3.2 hPa (20 °C)
Flash Point 44 °C
Explosion Limit 0.34 - 6.3 %(V)
Density 0.994 g/cm3 (20 °C)
pH (H2O, 20 °C) not applicable
Ignition Point 475 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR