Ethylbenzene zS, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Phenylethane

MQ200

8.01372.0100 8.01372.1000
8.01372.0100EA 23 GBP
8.01372.0100 8.01372.1000
Ethylbenzene zS, Sigma-Aldrich®
Ethylbenzene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 136.2
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 15
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
EINECS: 202-849-4
UN: 1175
ADR: 3 II
Merck Index: 13,03801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67d12bb66494ff09657a029fa1e68b09af34358&vid=3e5678dd6a0342c5ec35b93e48587e8125977cf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.2 g/l (20 °C)
Melting Point -95 °C
Molar Mass 106.16 g/mol
Boiling Point 136 °C (1013 hPa)
Vapor Pressure 9.5 hPa (20 °C)
Flash Point 15 °C
Explosion Limit 1.0 - 7.8 %(V)
Density 0.87 g/cm3 (20 °C)
Ignition Point 430 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR