Ethyl chloroformate for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Chloroformic acid ethyl ester

M-Clarity™ quality level = MQ200

8.00881.9025
8.00881.9025EA 0 GBP
8.00881.9025
Ethyl chloroformate for synthesis, Sigma-Aldrich®
Ethyl chloroformate
Formula: C₃H₅ClO₂
MW: 108.52 g/mol
Boiling Pt: 93
Melting Pt: –81 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 10
Storage Temperature: Ambient
MDL Number: MFCD00000644
CAS Number: 541-41-3
EINECS: 208-778-5
UN: 1182
ADR: 6.1 (3, 8) I
Merck Index: 12,03831

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR