Stearic acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Octadecanoic acid, Octadecylic acid

M-Clarity™ quality level = MQ200

8.00673.0500 8.00673.1000 8.00673.9050
8.00673.0500EA 26.8 GBP
8.00673.0500 8.00673.1000 8.00673.9050
Stearic acid for synthesis, Sigma-Aldrich®
Stearic acid
Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 361
Melting Pt: 67
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Molar Mass 284.49 g/mol
Storage Store at +15°C to +25°C
Grade REAG
Synonyms Octadecanoic Acid
Chemical Formula CH3(CH2)16CO2H


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR