Maleic anhydride for synthesis, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


M-Clarity™ quality level = MQ200

8.00408.0100 8.00408.0500 8.00408.1000 8.00408.5000 8.00408.9050 8.00408.9025
8.00408.0100EA 26.2 GBP
8.00408.0100 8.00408.0500 8.00408.1000 8.00408.5000 8.00408.9050 8.00408.9025
Maleic anhydride for synthesis, Sigma-Aldrich®
Maleic anhydride
Formula: C₄H₂O₃
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 51…53 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00005518
CAS Number: 108-31-6
EINECS: 203-571-6
UN: 2215
ADR: 8,III
Merck Index: 13,05727
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e48fcfd091622f49001009b383d7d62edf3521f5&vid=ca4d52f333124f172be23afe0d6384d67fc57a66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble,(decomposition)
Melting Point 51 - 53 °C
Molar Mass 98.06 g/mol
Bulk Density 700 - 800 kg/m3
Boiling Point 202 °C (1013 hPa)
Vapor Pressure 2.8 hPa (52.9 °C)
Flash Point 103 °C
Explosion Limit 1.4 - 7.1 %(V)
Density 1.32 g/cm3 (55 °C)
pH 0.8 (550 g/l, H2O, 20 °C) Hydrolysis
Ignition Point 475 °C DIN 51794
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR