Biotin kits, Micro HOOK™
Supplier: G Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-694 786-695 786-696 786-697
786-694EA 684 GBP
786-694 786-695 786-696 786-697
Biotin kits, Micro HOOK™
Protein Linking and Labeling Systems
The micro HOOK™ Biotin kits are designed to label small amounts of proteins, with each kit designed for 8 to 10 labellings of 50 to 250 µg protein/reaction.
  • Micro kit for labelling protein primary amines
  • Optimizer Buffer™ for improved coupling efficiency
  • Gel filtration columns for rapid (<10 minute) purification
  • Labels 50 to 250µg protein/reaction
  • Suitable for 8 to 10 couplings
Kits contain the following Biotin reagents
HOOK™ Sulfo-NHS-Biotin - amine reactive reagent, shortest spacer arm
HOOK™ Sulfo-NHS-LC-Biotin - amine reactive reagent, longer spacer arm
HOOK™ Sulfo-NHS-SS-Biotin - cleavable, amine reactive reagent
HOOK™ NHS-dPEG₄-Biotin - amine reactive pegylated reagent
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df83296ec1e7f955f878dcb6206fda945643b803&vid=dc7eee65ee2cea60a57858f6ad79f40fcf6f0d16&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d540f70135ec6fb5ea5eaf26687f26494e02f880&vid=993d813daacc154489875b5faa1b9d377deb3caa&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d6aef171e1842d10a195e754a9342cf1cde383a&vid=63fbc1af319e5f9a094438285c08ce441ee1efc8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59e20fd06123275c55f6d813d152a88f565191b7&vid=72ebb93b13ad8fafb69c35fde2ac8d65e6ed8ebb&prid=web&clid=SAPDAM