Washing solution, Protein-OUT™

Supplier: G Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

786-680
786-680EA 215 GBP
786-680
Washing solution, Protein-OUT™
Protein Extraction Protein Extraction Reagents

Protein-OUT™ is a washing solution that cleans plastic and glassware used in the proteolytic digestion and peptide isolation for mass spectrometry analysis. The rinsing of plastic and glassware with Protein-OUT™ will remove interfering chemicals and peptides that can leach from the tube walls and interfere or suppress the mass spectrometry signal. In addition, rinsing with Protein-OUT™ removes inadvertent contamination with dust and keratin.


  • Cost effective alternative to siliconised tubes
  • Supplied in 50 ml bottles
  • Suitable for the rinsing of ~200 1,5 - 2 ml micro centrifuge tubes

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR