Jacalin for IgA purification
Supplier: G Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-167 786-473
786-167EA 539 GBP
786-167 786-473
Jacalin for IgA purification
Protein Purification Protein Purification Reagents
Jacalin, or Artocarpus integrifolia lectin, is a tetrameric two-chain lectin with a molecular weight of 66 kDa. It is a α-D-galactose binding lectin purified from jack fruit (Artocarpus integrifolia) seeds.
Applications include isolating IgA from human serum and colostrums, isolating human plasma glycoproteins and histochemistry. It also binds IgD.