Transfer pads for western blotting, Swift™
Supplier: G Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-370 786-371 786-373 786-374 786-375 786-372
786-370EA 123 GBP
786-370 786-371 786-373 786-374 786-375 786-372
Transfer pads for western blotting, Swift™
Electrophoresis Reagents Western Blotting Reagents
SWIFT™ transfer pads are treated with a proprietary electrolyte buffer to enhance Western blot transfer efficiency.
  • High efficiency protein transfer
  • Reduce transfer time by up to 50%
  • No overheating or power shorts
  • No distortion or poor high molecular weight protein transfer