Western blot stripping buffer, Western ReProbe™
Supplier: G Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-119 786-306 786-305
786-119EA 278 GBP
786-119 786-306 786-305
Western blot stripping buffer, Western ReProbe™
Electrophoresis Reagents Western Blotting Reagents
Western ReProbe™ is a single component system that is specifically formulated to dissociate and remove antibodies from membrane-bound proteins without destroying the antigenic binding affinity. Western ReProbe™ also enables the ability to reuse Western blots.
  • Simply incubate the blot in Western ReProbe™ at room temperature and wash - no boiling, denaturants or SDS required
  • Repeated analysis of Western blots with multiple probes
  • Re-analysis and correction of unsatisfactory Western blots
  • Conservation of hard-to-obtain test samples and reagents
  • Time saving in not rerunning gels and transfer processes
Delivery information: Western ReProbe™ is supplied as a 5X concentrated solution. Each 100 ml is sufficient for 25 to 30 standard size (7,5×8,5 cm) Western blots.