Sample clean up, Perfect-FOCUS™
Supplier: G Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-124T 786-124
786-124TEA 93.3 GBP
786-124T 786-124
Sample clean up, Perfect-FOCUS™
Electrophoresis Reagents Western Blotting Reagents
Perfect-FOCUS™ is based on quantitative precipitation and concentration of protein solutions using UPPA™ (Universal Protein Precipitation Agent) and has been specifically developed for preparing lower conductivity protein samples for iso-electric focusing gels. Perfect-FOCUS™ is designed to clean and remove interfering agents from protein samples and concentrate protein solutions that give poor protein spot resolution during 2D electrophoresis.
Protein solutions as dilute as 1 ng/ml, if treated with Perfect-FOCUS™, can be quantitatively precipitated into a small volume. Protein precipitation is not affected by the presence of detergents or other common laboratory agents.
The collected protein sample has a conductivity of approximately 40 to 50 µS and is substantially free from non-protein agents, making the protein sample ideal for critical IEF and 2D gel electrophoresis studies, with improved spot resolution and greatly reduced spot streaking. Perfect-FOCUS™ treated protein samples are compatible with mass spectrometry analysis and show identical peptide spectra.