Gel loading dyes, Glow™
Supplier: G Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-100 786-101 786-102 786-103 786-104 786-105 786-106 786-107
786-100EA 85.7 GBP
786-100 786-101 786-102 786-103 786-104 786-105 786-106 786-107
Gel loading dyes, Glow™
Stains Electrophoresis Stains
These premixed and ready-to-use gel dyes are designed to minimise the use of ethidium bromide. Glow dyes provide intense DNA/RNA bands with little background or band distortion on any type of agarose or acrylamide gel.
  • Dyes for DNA are Ficoll®-based
  • Glow loading dyes already contain ethidium bromide
  • Both RNA loading dyes are formamide-based
  • Glow RNA for quick screenings of RNA preps
Specifically formulated for nucleic acid electrophoresis.