Membranes and padding for Western protein transfers, pre-cut
Supplier: G-Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-018NC 786-018PV 786-056PV
786-018NCEA 351 GBP
786-018NC 786-018PV 786-056PV
Membranes and padding for Western protein transfers, pre-cut
Transfer Membranes
Pre-cut transfer membranes and padding for Western blot transfer procedures. Pre-cut membranes are supplied sandwiched between blotting paper padding. Simply soak the membrane in transfer buffer and assemble with the gel in a transfer cassette. Nitrocellulose (0,22 µm pore size) and PVDF (0,45 µm pore size) membranes are available.