Titanium(IV) isopropoxide 97

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Titanium tetraisopropanolate, Titanium (IV) isopropoxide

77115.09 77115.22 77115.36 77115.57
77115.09EA 26.3 GBP
77115.09 77115.22 77115.36 77115.57
Titanium(IV) isopropoxide 97
Titanium (IV) isopropoxide

Purity: 97+% CAS No.: 546-68-9 Molecular Formula: C12H28O4Ti Formula Weight: 284.23 Flash Point: 46°(115°F) Physical Form: Liquid Density: 0.955 Refractive Index: 1.4640 Sensitivity: Moisture Sensitive Appearance: Colourless to pale yellow Melting Point: 16-20° Boiling Point: 232° Solubility: Soluble in anhydrous ethanol, ether, benzene and chloroform. Decomposes in water Beilstein Reference: 3679474 Merck Reference: 14,9480 UN No.: 2413 MDL No.: MFCD00008871


Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids

Formula: C₁₂H₂₈O₄Ti
MW: 284.23 g/mol
Boiling Pt: 232 °C
Melting Pt: 16…20 °C
Density: 0.955
Flash Pt: 46 °C (115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008871
CAS Number: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3 III
Merck Index: 12,09614

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR