Flow cups with fixed nozzle
Supplier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VF2048 VF2183 VF2049 VF2051 VF2050
769-1152EA 210 GBP
769-1152 SIMEVF2183 SIMEVF2049 SIMEVF2051 SIMEVF2050
Flow cups with fixed nozzle
Viscometers Flow Cup Viscometers
These flow cups are composed of an anodised aluminium cup and a stainless steel nozzle.
Designed to measure the viscosity of lacquers, paint and other liquids.
Certifications: ISO 2431
Ordering information: Stand must be ordered separately.