n-Pentane ≥99.0% (by GC), CHROMASOLV™ for residue analysis (of high-volatile halogenated hydrocarbons), Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Pentane

76866-1L
76866-1LEA 139 GBP
76866-1L
n-Pentane ≥99.0% (by GC), CHROMASOLV™ for residue analysis (of high-volatile halogenated hydrocarbons), Riedel-de Haën™
n-Pentane

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36…37 °C
Melting Pt: –131 °C
Density: 0.631 g/cm³ (15 °C)
Flash Pt: <–35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Explosion Limit ~8.3 %
Assay (GC) Min. 99 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Suitability for halogen hydrocarbons Max. 5 µg/l
Refractive Index n20/D 1.358 (lit.)
Autoignition Temperature 500 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR